Flavin adenine dinucleotide disodium salt hydrate ≥98%

Supplier: Ambeed
Warning

A1207949-100MG A1207949-250MG A1207949-1G
77596-260EA 58.43 USD
77596-260 77620-840 77866-621
Flavin adenine dinucleotide disodium salt hydrate ≥98%
Flavin adenine dinucleotide disodium salt hydrate

Precaución: for Research Use Only

Formula: C₂₇H₃₃N₉Na₂O₁₆P₂
MDL Number: MFCD00151217
CAS Number: 1891059-93-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR