Flavin adenine dinucleotide disodium salt ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl)methoxy)(((((2R,3S,4S)-5-(7,8-dimethyl-2,4-dioxo-2H,3H,4H,10H-benzo[g]pteridin-10-yl)-2,3,4-trihydroxypentyl)oxy)(hydroxy)phosphoryl)oxy)phosphinic acid Disodium Salt, FAD-Na2, Flavin Adenine Dinucleotide Disodium Salt, Riboflavin 5'-adenosine diphosphate disodium salt, FAD Disodium Salt
A1207948-100MG
A1207948-1G
A1207948-250MG
A1207948-5G
77596-254EA
120.95
USD
77596-254
77596-256
77596-258
77866-620
Flavin adenine dinucleotide disodium salt ≥98%
Flavin adenine dinucleotide disodium salt
Precaución: for Research Use Only
Formula:
C₂₇H₃₃N₉O₁₅P₂ MW: 829.52 g/mol |
MDL Number:
MFCD00151217 CAS Number: 84366-81-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...