2-Bromo-2'-methoxy-1,1'-biphenyl ≥98%

Supplier: Ambeed
Warning

A1206125-100MG A1206125-1G
77866-585EA 43 USD
77866-585 77866-586
2-Bromo-2'-methoxy-1,1'-biphenyl ≥98%
2-Bromo-2'-methoxy-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₃H₁₁BrO
MDL Number: MFCD16659071
CAS Number: 20837-12-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR