9,9-Dimethyl-9H-fluoren-3-amine ≥98%

Supplier: Ambeed
Warning

A1206122-1G A1206122-250MG
77620-812EA 34.19 USD
77620-812 77866-584
9,9-Dimethyl-9H-fluoren-3-amine ≥98%
9,9-Dimethyl-9H-fluoren-3-amine

Precaución: for Research Use Only

Formula: C₁₅H₁₅N
MDL Number: MFCD31385005
CAS Number: 1421789-14-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR