(S)-6-(4-Fluorophenoxy)-3-((1-isopropylpiperidin-3-yl)methyl)-2-methylquinazolin-4(3H)-one ≥97%

Supplier: Ambeed
Warning

Synonyms: YIL781

A1206036-1MG A1206036-10MG A1206036-5MG
77596-242EA 46.13 USD
77596-242 77620-808 77620-810
(S)-6-(4-Fluorophenoxy)-3-((1-isopropylpiperidin-3-yl)methyl)-2-methylquinazolin-4(3H)-one ≥97%
(S)-6-(4-Fluorophenoxy)-3-((1-isopropylpiperidin-3-yl)methyl)-2-methylquinazolin-4(3H)-one

Precaución: for Research Use Only

Formula: C₂₄H₂₈FN₃O₂
MW: 409.5 g/mol
MDL Number: MFCD27980545
CAS Number: 875258-85-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR