2-Methoxy-3(or 5)-methylpyrazine 98% mix

Supplier: Ambeed
Danger

A1205941-5G A1205941-25G
77620-806EA 6.18 USD
77620-806 77866-580
2-Methoxy-3(or 5)-methylpyrazine 98% mix
2-Methoxy-3(or 5)-methylpyrazine

Precaución: for Research Use Only

Formula: C₆H₈N₂O
CAS Number: 68378-13-2
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR