Methyl 3-chloro-4-hydroxy-5-nitrobenzoate ≥98%

Supplier: Ambeed
Danger

A1197642-1G A1197642-5G
77620-790EA 30.9 USD
77620-790 77620-792
Methyl 3-chloro-4-hydroxy-5-nitrobenzoate ≥98%
Methyl 3-chloro-4-hydroxy-5-nitrobenzoate

Precaución: for Research Use Only

Formula: C₈H₆ClNO₅
MDL Number: MFCD27950418
CAS Number: 40256-81-3
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR