2,2'-(7-(2-((2-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)ethyl)amino)-2-oxoethyl)-1,4,7-triazonane-1,4-diyl)diacetic acid ≥98%

Supplier: Ambeed

Synonyms: Maleimide-NOTA

A1197477-100MG A1197477-25MG A1197477-50MG
77866-558EA 389 USD
77866-558 77866-559 77866-560
2,2'-(7-(2-((2-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)ethyl)amino)-2-oxoethyl)-1,4,7-triazonane-1,4-diyl)diacetic acid ≥98%
2,2'-(7-(2-((2-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)ethyl)amino)-2-oxoethyl)-1,4,7-triazonane-1,4-diyl)diacetic acid

Precaución: for Research Use Only

Formula: C₁₈H₂₇N₅O₇
CAS Number: 1295584-83-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR