4-Methylpyrrolidin-2-one ≥98%

Supplier: Ambeed
Warning

A119744-1G A119744-5G
77592-332EA 79.95 USD
77592-332 77889-829
4-Methylpyrrolidin-2-one ≥98%
4-Methylpyrrolidin-2-one

Precaución: for Research Use Only

Formula: C₅H₉NO
MDL Number: MFCD00128875
CAS Number: 2996-58-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR