3-Bromo-4-iodo-1,1'-biphenyl ≥97%

Supplier: Ambeed
Danger

A1196815-1G A1196815-250MG
77571-178EA 32 USD
77571-178 77866-555
3-Bromo-4-iodo-1,1'-biphenyl ≥97%
3-Bromo-4-iodo-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₂H₈BrI
MDL Number: MFCD28952954
CAS Number: 900806-53-3
UN: 3152
ADR: 9,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR