6-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)-N-((S)-1-(((S)-1-((4-(hydroxymethyl)phenyl)amino)-1-oxopropan-2-yl)amino)-3-methyl-1-oxobutan-2-yl)hexanamide ≥98%

Supplier: Ambeed
Warning

Synonyms: Mc-Val-Ala-PAB

A1196787-1G A1196787-100MG A1196787-250MG
77571-176EA 742.63 USD
77571-176 77620-786 77620-788
6-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)-N-((S)-1-(((S)-1-((4-(hydroxymethyl)phenyl)amino)-1-oxopropan-2-yl)amino)-3-methyl-1-oxobutan-2-yl)hexanamide ≥98%
6-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)-N-((S)-1-(((S)-1-((4-(hydroxymethyl)phenyl)amino)-1-oxopropan-2-yl)amino)-3-methyl-1-oxobutan-2-yl)hexanamide

Precaución: for Research Use Only

Formula: C₂₅H₃₄N₄O₆
MW: 486.5607 g/mol
MDL Number: MFCD32063500
CAS Number: 1870916-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR