O-Phospho-L-tyrosine ≥98%

Supplier: Ambeed
Warning

Synonyms: H-tyr(H2po3)-oh, H-Tyr(PO₃H₂)-OH, H-Tyr(H₂PO₃)-OH

A119618-1G A119618-5G A119618-250MG
77599-722EA 73.8 USD
77599-722 77599-724 77666-706
O-Phospho-L-tyrosine ≥98%
O-Phospho-L-tyrosine

Precaución: for Research Use Only

Formula: C₉H₁₂NO₆
MW: 261.17 g/mol
MDL Number: MFCD00002603
CAS Number: 21820-51-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR