4,4',4'',4''',4'''',4'''''-(9,10-Dihydro-9,10-[1,2]benzenoanthracene-2,3,6,7,14,15-hexayl)hexabenzoic acid ≥98%

Supplier: Ambeed
Warning

A1195919-1G A1195919-100MG
77620-776EA 221.45 USD
77620-776 77866-542
4,4',4'',4''',4'''',4'''''-(9,10-Dihydro-9,10-[1,2]benzenoanthracene-2,3,6,7,14,15-hexayl)hexabenzoic acid ≥98%
4,4',4'',4''',4'''',4'''''-(9,10-Dihydro-9,10-[1,2]benzenoanthracene-2,3,6,7,14,15-hexayl)hexabenzoic acid

Precaución: for Research Use Only

Formula: C₆₂H₃₈O₁₂
MDL Number: MFCD34165526
CAS Number: 1835723-11-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR