1,3,5-Triiodo-2,4,6-trimethylbenzene ≥98%

Supplier: Ambeed
Warning

A1195910-5G A1195910-1G A1195910-250MG
77571-170EA 173 USD
77571-170 77620-774 77866-540
1,3,5-Triiodo-2,4,6-trimethylbenzene ≥98%
1,3,5-Triiodo-2,4,6-trimethylbenzene

Precaución: for Research Use Only

Formula: C₉H₉I₃
CAS Number: 19025-36-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR