5,10,15,20-Tetrakis(4-(1,3,2-dioxaborinan-2-yl)phenyl)porphyrin ≥98%

Supplier: Ambeed
Warning

A1195895-1G A1195895-250MG
77620-770EA 246.17 USD
77620-770 77620-772
5,10,15,20-Tetrakis(4-(1,3,2-dioxaborinan-2-yl)phenyl)porphyrin ≥98%
5,10,15,20-Tetrakis(4-(1,3,2-dioxaborinan-2-yl)phenyl)porphyrin

Precaución: for Research Use Only

Formula: C₅₆H₅₀B₄N₄O₈
MW: 950.2622 g/mol
CAS Number: 1270214-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR