(T-4)-[3-BRomo-5-[(4-bromo-3,5-dimethyl-2H-pyrrol-2-ylidene-κN)phenylmethyl]-2,4-dimethyl-1H-pyrrolato-κN]difluoroboron ≥95%

Supplier: Ambeed
Warning

A1195875-1G A1195875-100MG A1195875-250MG A1195875-5G
77592-326EA 330.05 USD
77592-326 77620-766 77620-768 77866-538
(T-4)-[3-BRomo-5-[(4-bromo-3,5-dimethyl-2H-pyrrol-2-ylidene-κN)phenylmethyl]-2,4-dimethyl-1H-pyrrolato-κN]difluoroboron ≥95%
(T-4)-[3-BRomo-5-[(4-bromo-3,5-dimethyl-2H-pyrrol-2-ylidene-κN)phenylmethyl]-2,4-dimethyl-1H-pyrrolato-κN]difluoroboron

Precaución: for Research Use Only

CAS Number: 910823-84-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR