2,5-Bis(dodecyloxy)terephthalaldehyde ≥98%

Supplier: Ambeed
Warning

A1195789-1G A1195789-100MG
77620-754EA 198.79 USD
77620-754 77866-528
2,5-Bis(dodecyloxy)terephthalaldehyde ≥98%
2,5-Bis(dodecyloxy)terephthalaldehyde

Precaución: for Research Use Only

Formula: C₃₂H₅₄O₄
CAS Number: 123415-45-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR