(S)-2-Amino-2-(2-fluorophenyl)acetic acid hydrochloride ≥97%

Supplier: Ambeed
Warning

A1195616-100MG A1195616-50MG
77866-519EA 129 USD
77866-519 77866-520
(S)-2-Amino-2-(2-fluorophenyl)acetic acid hydrochloride ≥97%
(S)-2-Amino-2-(2-fluorophenyl)acetic acid hydrochloride

Precaución: for Research Use Only

Formula: C₈H₉ClFNO₂
MDL Number: MFCD12759025
CAS Number: 2829279-60-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR