21-Hydroxyhenicosanoic acid ≥98%

Supplier: Ambeed
Warning

A1195099-1G A1195099-250MG A1195099-5G
77571-162EA 80 USD
77571-162 77596-228 77596-230
21-Hydroxyhenicosanoic acid ≥98%

Precaución: for Research Use Only

Formula: C₂₁H₄₂O₃
MDL Number: MFCD02259051
CAS Number: 89160-04-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR