tert-Butyl 6-oxohexanoate ≥95%

Supplier: Ambeed
Warning

A1195068-100MG A1195068-250MG
77596-226EA 101.48 USD
77596-226 77620-744
tert-Butyl 6-oxohexanoate ≥95%
tert-Butyl 6-oxohexanoate

Precaución: for Research Use Only

Formula: C₁₀H₁₈O₃
MW: 186.2481 g/mol
CAS Number: 84201-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR