(4-Methyl-1H-pyrazol-3-yl)boronic acid ≥98%

Supplier: Ambeed

A1193273-100MG A1193273-1G
77783-331EA 175 USD
77783-331 77783-332
(4-Methyl-1H-pyrazol-3-yl)boronic acid ≥98%
(4-Methyl-1H-pyrazol-3-yl)boronic acid

Precaución: for Research Use Only

Formula: C₄H₇BN₂O₂
MDL Number: MFCD31721373
CAS Number: 1562244-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR