4,4'-Methylenebis(N-(sec-butyl)aniline) ≥97%

Supplier: Ambeed
Danger

A1192830-1000G A1192830-100G A1192830-25G
77571-146EA 109 USD
77571-146 77866-484 77866-485
4,4'-Methylenebis(N-(sec-butyl)aniline) ≥97%
4,4'-Methylenebis(N-(sec-butyl)aniline)

Precaución: for Research Use Only

Formula: C₂₁H₃₀N₂
MDL Number: MFCD03703459
CAS Number: 5285-60-9
UN: 3082
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR