3',5'-Difluoroacetophenone ≥98%

Supplier: Ambeed
Warning

Synonyms: 1-(3,5-Difluorophenyl)ethan-1-one

A119252-10G A119252-1G A119252-5G
77685-049EA 15 USD
77685-049 77685-050 77685-051
3',5'-Difluoroacetophenone ≥98%
3',5'-Difluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₆F₂O
MW: 156.13 g/mol
MDL Number: MFCD00042489
CAS Number: 123577-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR