3,5-Dibromo-2,6-difluorobenzonitrile ≥97%

Supplier: Ambeed
Danger

A1191869-1G A1191869-100MG A1191869-250MG
77620-710EA 79.31 USD
77620-710 77866-468 77866-469
3,5-Dibromo-2,6-difluorobenzonitrile ≥97%
3,5-Dibromo-2,6-difluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇HBr₂F₂N
MW: 296.8944 g/mol
MDL Number: MFCD28783426
CAS Number: 1806299-03-5
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR