(2-Methyl-6-(trifluoromethyl)phenyl)boronic acid 98+%

Supplier: Ambeed

A119046-100MG A119046-1G A119046-250MG
77885-082EA 175 USD
77885-082 77885-083 77885-084
(2-Methyl-6-(trifluoromethyl)phenyl)boronic acid 98+%
(2-Methyl-6-(trifluoromethyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₈BF₃O₂
CAS Number: 2305931-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR