Ethyl 1,3,4-thiadiazole-2-carboxylate ≥98%

Supplier: Ambeed
Warning

A118952-100MG A118952-250MG
77670-678EA 58.6 USD
77670-678 77882-970
Ethyl 1,3,4-thiadiazole-2-carboxylate ≥98%
Ethyl 1,3,4-thiadiazole-2-carboxylate

Precaución: for Research Use Only

Formula: C₅H₆N₂O₂S
MDL Number: MFCD14155685
CAS Number: 1378819-08-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR