(R)-3,3'-Dimesityl-5,5',6,6',7,7',8,8'-octahydro-[1,1'-binaphthalene]-2,2'-diol ≥98%

Supplier: Ambeed
Warning

A1188924-100MG A1188924-250MG A1188924-1G
77596-204EA 58.43 USD
77596-204 77596-206 77620-694
(R)-3,3'-Dimesityl-5,5',6,6',7,7',8,8'-octahydro-[1,1'-binaphthalene]-2,2'-diol ≥98%
(R)-3,3'-Dimesityl-5,5',6,6',7,7',8,8'-octahydro-[1,1'-binaphthalene]-2,2'-diol

Precaución: for Research Use Only

CAS Number: 400027-51-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR