(E)-3-(3,5-di-tert-butyl-4-hydroxybenzylidene)dihydrofuran-2(3H)-one ≥98%

Supplier: Ambeed
Warning

Synonyms: PGS-IN-1

A1188681-1MG A1188681-10MG A1188681-5MG
77596-202EA 184.5 USD
77596-202 77620-690 77620-692
(E)-3-(3,5-di-tert-butyl-4-hydroxybenzylidene)dihydrofuran-2(3H)-one ≥98%
(E)-3-(3,5-di-tert-butyl-4-hydroxybenzylidene)dihydrofuran-2(3H)-one

Precaución: for Research Use Only

Formula: C₁₉H₂₆O₃
MW: 302.41 g/mol
MDL Number: MFCD30747896
CAS Number: 102271-49-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR