4-Bromo-6-nitro-1H-indazole ≥95%

Supplier: Ambeed
Warning

A118805-1G A118805-100MG A118805-250MG
77610-980EA 66.95 USD
77610-980 77670-668 77670-670
4-Bromo-6-nitro-1H-indazole ≥95%
4-Bromo-6-nitro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄BrN₃O₂
MW: 242.03 g/mol
MDL Number: MFCD07781323
CAS Number: 885518-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR