1,1'-Dimethyl-[2,2'-bipyridine]-1,1'-diium iodide ≥98%

Supplier: Ambeed
Warning

A1188037-1G A1188037-250MG A1188037-5G
77620-676EA 50.47 USD
77620-676 77866-441 77866-442
1,1'-Dimethyl-[2,2'-bipyridine]-1,1'-diium iodide ≥98%
1,1'-Dimethyl-[2,2'-bipyridine]-1,1'-diium iodide

Precaución: for Research Use Only

Formula: C₁₂H₁₄I₂N₂
MDL Number: MFCD02751858
CAS Number: 23484-62-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR