3,3'-((2-(((Benzyloxy)carbonyl)amino)-2-((2-carboxyethoxy)methyl)propane-1,3-diyl)bis(oxy))dipropionic acid ≥95%

Supplier: Ambeed
Warning

A1188012-250MG A1188012-100MG A1188012-10G
77592-312EA 104.55 USD
77592-312 77596-192 77620-674
3,3'-((2-(((Benzyloxy)carbonyl)amino)-2-((2-carboxyethoxy)methyl)propane-1,3-diyl)bis(oxy))dipropionic acid ≥95%
3,3'-((2-(((Benzyloxy)carbonyl)amino)-2-((2-carboxyethoxy)methyl)propane-1,3-diyl)bis(oxy))dipropionic acid

Precaución: for Research Use Only

Formula: C₂₁H₂₉NO₁₁
MW: 471.46 g/mol
MDL Number: MFCD28966608
CAS Number: 200133-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR