(1-(Methylsulfonyl)-1H-pyrazol-4-yl)boronic acid ≥98%

Supplier: Ambeed

A1188002-1G A1188002-250MG
77571-116EA 180 USD
77571-116 77866-439
(1-(Methylsulfonyl)-1H-pyrazol-4-yl)boronic acid ≥98%
(1-(Methylsulfonyl)-1H-pyrazol-4-yl)boronic acid

Precaución: for Research Use Only

Formula: C₄H₇BN₂O₄S
MDL Number: MFCD18427609
CAS Number: 1201643-87-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR