(S)-2-Amino-2-(3,5-dimethylphenyl)ethan-1-ol ≥95%

Supplier: Ambeed
Warning

A1186619-100MG A1186619-1G
77866-420EA 142 USD
77866-420 77866-421
(S)-2-Amino-2-(3,5-dimethylphenyl)ethan-1-ol ≥95%
(S)-2-Amino-2-(3,5-dimethylphenyl)ethan-1-ol

Precaución: for Research Use Only

Formula: C₁₀H₁₅NO
MDL Number: MFCD09253999
CAS Number: 1213317-54-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR