(R)-1-(4-Chloro-2-fluorophenyl)ethan-1-amine hydrochloride ≥95%

Supplier: Ambeed
Warning

A1186589-250MG A1186589-100MG A1186589-1G
77592-308EA 203.98 USD
77592-308 77620-656 77620-658
(R)-1-(4-Chloro-2-fluorophenyl)ethan-1-amine hydrochloride ≥95%
(R)-1-(4-Chloro-2-fluorophenyl)ethan-1-amine hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₀Cl₂FN
MDL Number: MFCD24414699
CAS Number: 856562-91-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR