1-(6-Chloropyridin-3-yl)cyclopropanecarbonitrile ≥95%

Supplier: Ambeed
Warning

A118552-100MG A118552-1G A118552-250MG
77891-847EA 12 USD
77891-847 77891-848 77891-849
1-(6-Chloropyridin-3-yl)cyclopropanecarbonitrile ≥95%
1-(6-Chloropyridin-3-yl)cyclopropanecarbonitrile

Precaución: for Research Use Only

Formula: C₉H₇ClN₂
MDL Number: MFCD15523665
CAS Number: 854267-89-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR