(1-((2-(Trimethylsilyl)ethoxy)methyl)-1H-pyrazol-4-yl)boronic acid ≥98%

Supplier: Ambeed

A1182484-100MG A1182484-1G
77866-413EA 45 USD
77866-413 77866-414
(1-((2-(Trimethylsilyl)ethoxy)methyl)-1H-pyrazol-4-yl)boronic acid ≥98%
(1-((2-(Trimethylsilyl)ethoxy)methyl)-1H-pyrazol-4-yl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₉BN₂O₃Si
MDL Number: MFCD31802301
CAS Number: 719296-53-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR