2-(Prop-2-yn-1-yloxy)ethyl 4-methylbenzenesulfonate ≥95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Propargyl-PEG2-Tos
A1181809-5G
A1181809-1G
A1181809-250MG
77596-184EA
342.35
USD
77596-184
77620-650
77620-652
2-(Prop-2-yn-1-yloxy)ethyl 4-methylbenzenesulfonate ≥95%
2-(Prop-2-yn-1-yloxy)ethyl 4-methylbenzenesulfonate
Precaución: for Research Use Only
Formula:
C₁₂H₁₄O₄S MW: 254.3022 g/mol |
MDL Number:
MFCD29764327 CAS Number: 145916-41-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...