1-Bromo-5-fluoronaphthalene ≥97%, Ambeed.Inc

Supplier: Ambeed
Warning

A118168-5G A118168-1G A118168-250MG
77683-064EA 367 USD
77683-064 77683-062 77683-063
1-Bromo-5-fluoronaphthalene ≥97%, Ambeed.Inc
1-Bromo-5-fluoronaphthalene

Precaución: For research use only.

Formula: C₁₀H₆BrF
MW: 225.0571 g/mol
MDL Number: MFCD09258916
CAS Number: 315-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR