2-(4-Fluoro-2-methylphenoxy)-N-(2-oxo-1,2-dihydropyridin-4-yl)-4-(trifluoromethyl)benzamide ≥95%

Supplier: Ambeed
Warning

Synonyms: Nav1.8-IN-4

A1181590-10MG A1181590-5MG A1181590-100MG A1181590-250MG A1181590-25MG A1181590-50MG
77620-642EA 185.08 USD
77620-642 77620-644 77866-398 77866-399 77866-400 77866-401
2-(4-Fluoro-2-methylphenoxy)-N-(2-oxo-1,2-dihydropyridin-4-yl)-4-(trifluoromethyl)benzamide ≥95%
2-(4-Fluoro-2-methylphenoxy)-N-(2-oxo-1,2-dihydropyridin-4-yl)-4-(trifluoromethyl)benzamide

Precaución: for Research Use Only

Formula: C₂₀H₁₄F₄N₂O₃
MDL Number: MFCD31803782
CAS Number: 1620846-16-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR