5-(2-Methyloctan-2-yl)benzene-1,3-diol ≥98%

Supplier: Ambeed
Warning

A117978-5G A117978-1G
77747-857EA 119 USD
77747-857 77747-856
5-(2-Methyloctan-2-yl)benzene-1,3-diol ≥98%
5-(2-Methyloctan-2-yl)benzene-1,3-diol

Precaución: for Research Use Only

Formula: C₁₅H₂₄O₂
MDL Number: MFCD03001733
CAS Number: 56469-10-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR