4-Amino-3-bromoquinoline ≥95%

Supplier: Ambeed
Warning

A117922-1G A117922-5G
77891-840EA 55 USD
77891-840 77891-841
4-Amino-3-bromoquinoline ≥95%
4-Amino-3-bromoquinoline

Precaución: for Research Use Only

Formula: C₉H₇BrN₂
MDL Number: MFCD00234502
CAS Number: 36825-36-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR