(4-(N,N-Dimethylsulfamoyl)-3-methylphenyl)boronic acid ≥98%

Supplier: Ambeed
Warning

A1178704-1G A1178704-250MG A1178704-100MG
77596-166EA 213.2 USD
77596-166 77596-168 77620-618
(4-(N,N-Dimethylsulfamoyl)-3-methylphenyl)boronic acid ≥98%
(4-(N,N-Dimethylsulfamoyl)-3-methylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₄BNO₄S
MW: 243.09 g/mol
MDL Number: MFCD31721010
CAS Number: 1778667-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR