Ethyl 3-(benzyloxy)-4-oxo-4H-pyran-2-carboxylate ≥98%

Supplier: Ambeed
Warning

A1178126-250MG A1178126-100MG
77620-616EA 37.08 USD
77620-616 77620-614
Ethyl 3-(benzyloxy)-4-oxo-4H-pyran-2-carboxylate ≥98%
Ethyl 3-(benzyloxy)-4-oxo-4H-pyran-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₅
MW: 274.2687 g/mol
MDL Number: MFCD31697637
CAS Number: 1332855-94-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR