2-(2-(2-Chloroethoxy)ethoxy)acetic acid ≥98%

Supplier: Ambeed
Danger

Synonyms: Cl-PEG2-acid

A1178106-100MG A1178106-1G A1178106-250MG A1178106-5G
77620-608EA 50.47 USD
77620-608 77620-610 77620-612 77866-370
2-(2-(2-Chloroethoxy)ethoxy)acetic acid ≥98%
2-(2-(2-Chloroethoxy)ethoxy)acetic acid

Precaución: for Research Use Only

Formula: C₆H₁₁ClO₄
MDL Number: MFCD20623441
CAS Number: 170304-76-4
UN: 2810
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR