(11bR)-2,6-Bis[3,5-bis(1,1-dimethylethyl)-4-methoxyphenyl]-4-hydroxy-4-oxide-dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin ≥97%

Supplier: Ambeed
Warning

A1177901-100MG A1177901-250MG
77596-162EA 200.9 USD
77596-162 77620-606
(11bR)-2,6-Bis[3,5-bis(1,1-dimethylethyl)-4-methoxyphenyl]-4-hydroxy-4-oxide-dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin ≥97%
(11bR)-2,6-Bis[3,5-bis(1,1-dimethylethyl)-4-methoxyphenyl]-4-hydroxy-4-oxide-dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin

Precaución: for Research Use Only

Formula: C₅₀H₅₇O₆P
MW: 784.96 g/mol
MDL Number: MFCD31747023
CAS Number: 957790-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR