N-(2,3-Dihydro-1H-inden-2-yl)-2,2-difluorobenzo[d][1,3]dioxole-5-carboxamide ≥99%

Supplier: Ambeed

A1177345-10MG A1177345-1MG A1177345-5MG
77596-148EA 101.47 USD
77596-148 77596-150 77596-152
N-(2,3-Dihydro-1H-inden-2-yl)-2,2-difluorobenzo[d][1,3]dioxole-5-carboxamide ≥99%
N-(2,3-Dihydro-1H-inden-2-yl)-2,2-difluorobenzo[d][1,3]dioxole-5-carboxamide

Precaución: for Research Use Only

Formula: C₁₇H₁₃F₂NO₃
MDL Number: MFCD30532601
CAS Number: 450348-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR