SB-204490 ≥98%

Supplier: Ambeed
Warning

Synonyms: 2-((3S,5R)-5-(6-(2,4-Dichlorophenyl)hexyl)-3-hydroxy-2-oxotetrahydrofuran-3-yl)acetic acid, SB 204990

A1177275-10MG A1177275-5MG
77866-357EA 147 USD
77866-357 77866-358
SB-204490 ≥98%
SB-204490

Precaución: for Research Use Only

Formula: C₁₈H₂₂Cl₂O₅
MDL Number: MFCD28127018
CAS Number: 154566-12-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR