7-Benzyl-2,3-dihydroxy-6-methyl-4-propyl-1-naphthoic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: FX-11

A1177274-10MG A1177274-5MG
77620-590EA 147.29 USD
77620-590 77620-592
7-Benzyl-2,3-dihydroxy-6-methyl-4-propyl-1-naphthoic acid ≥98%
7-Benzyl-2,3-dihydroxy-6-methyl-4-propyl-1-naphthoic acid

Precaución: for Research Use Only

Formula: C₂₂H₂₂O₄
MW: 350.41 g/mol
MDL Number: MFCD22572764
CAS Number: 213971-34-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR