1-((1-(2-Aminopyrimidin-4-yl)-2-((2-methoxyethyl)amino)-1H-benzo[d]imidazol-6-yl)ethynyl)cyclohexan-1-ol ≥99%

Supplier: Ambeed
Warning

Synonyms: GNE 2861

A1177174-1MG A1177174-5MG
77596-128EA 35.88 USD
77596-128 77620-580
1-((1-(2-Aminopyrimidin-4-yl)-2-((2-methoxyethyl)amino)-1H-benzo[d]imidazol-6-yl)ethynyl)cyclohexan-1-ol ≥99%
1-((1-(2-Aminopyrimidin-4-yl)-2-((2-methoxyethyl)amino)-1H-benzo[d]imidazol-6-yl)ethynyl)cyclohexan-1-ol

Precaución: for Research Use Only

Formula: C₂₂H₂₆N₆O₂
MW: 406.48 g/mol
MDL Number: MFCD28399125
CAS Number: 1394121-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR