8-Chloro-2-((2,4-dichlorophenyl)amino)-3-(3-methylbutanoyl)-5-nitroquinolin-4(1H)-one ≥99%

Supplier: Ambeed

A1177170-10MG A1177170-1MG A1177170-5MG
77596-122EA 742.1 USD
77596-122 77596-124 77596-126
8-Chloro-2-((2,4-dichlorophenyl)amino)-3-(3-methylbutanoyl)-5-nitroquinolin-4(1H)-one ≥99%
8-Chloro-2-((2,4-dichlorophenyl)amino)-3-(3-methylbutanoyl)-5-nitroquinolin-4(1H)-one

Precaución: for Research Use Only

Formula: C₂₀H₁₆Cl₃N₃O₄
MDL Number: MFCD32197217
CAS Number: 1361227-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR